Assay Details
| Gene Symbol: LOC4326931 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765833.2 | XP_015621319.1 | 2 - 3 | 994 | 98 | |
| GenBank mRNA | AK066793.1 | - | 2 - 3 | 573 | 98 | |
More Information
Set Membership: |
|

LOC4326931
Genomic Map