Assay Details
Gene Symbol: LOC4327014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015779017.2 | XP_015634503.1 | 5 - 6 | 1174 | 93 | |
GenBank mRNA | AK072330.1 | - | 5 - 6 | 1132 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
