Assay Details
| Gene Symbol: LOC4327593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770951.2 | 6 - 7 | 691 | 68 | ||
| GenBank mRNA | AK063065.1 | - | 1 - 2 | 71 | 68 | |
| AK067934.1 | - | 5 - 6 | 651 | 68 | ||
More Information
Set Membership: |
|

LOC4327593
Genomic Map