Assay Details
| Gene Symbol: LOC4327593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770951.2 | 7 - 8 | 946 | 92 | ||
| GenBank mRNA | AK063065.1 | - | 2 - 3 | 326 | 92 | |
| AK067934.1 | - | 6 - 7 | 906 | 92 | ||
More Information
Set Membership: |
|

LOC4327593
Genomic Map