Assay Details
| Gene Symbol: LOC4327493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757505.2 | XP_015612991.1 | 2 - 3 | 322 | 69 | |
| GenBank mRNA | AK060836.1 | - | 1 - 2 | 172 | 69 | |
| AK065059.1 | - | 2 - 3 | 260 | 69 | ||
| KC140120.1 | - | 2 - 3 | 251 | 69 | ||
More Information
Set Membership: |
|

LOC4327493
Genomic Map