Assay Details
Gene Symbol: LOC4327615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766602.2 | XP_015622088.1 | 1 - 2 | 264 | 61 | |
GenBank mRNA | AK103249.1 | - | 1 - 2 | 205 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015766602.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
