Assay Details
Gene Symbol: LOC4327615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766601.2 | XP_015622087.1 | 3 - 4 | 1197 | 109 | |
XM_015766602.2 | XP_015622088.1 | 3 - 4 | 1179 | 109 | ||
GenBank mRNA | AK103249.1 | - | 3 - 4 | 1120 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
