Assay Details
Gene Symbol: LOC107275263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766141.2 | XP_015621627.1 | 1 - 2 | 556 | 65 | |
XM_015766142.2 | XP_015621628.1 | 1 - 2 | 556 | 65 | ||
XM_015766144.2 | XP_015621630.1 | 1 - 2 | 556 | 65 | ||
GenBank mRNA | AK103032.1 | - | 1 - 2 | 310 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015766141.2, XM_015766142.2, XM_015766144.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
