Assay Details
| Gene Symbol: LOC9267360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765535.2 | XP_015621021.1 | 10 - 11 | 1335 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC9267360| Gene Symbol: LOC9267360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765535.2 | XP_015621021.1 | 10 - 11 | 1335 | 101 | |
Set Membership: |
|