Assay Details
| Gene Symbol: LOC4324891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778363.2 | XP_015633849.1 | 1 - 2 | 775 | 106 | |
| GenBank mRNA | AK070098.1 | - | 1 - 2 | 369 | 106 | |
More Information
Set Membership: |
|

LOC4324891
Genomic Map