Assay Details
| Gene Symbol: LOC4324891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778363.2 | XP_015633849.1 | 7 - 8 | 1221 | 81 | |
| XM_015778432.2 | XP_015633918.1 | 8 - 9 | 636 | 81 | ||
| GenBank mRNA | AK070098.1 | - | 7 - 8 | 815 | 81 | |
More Information
Set Membership: |
|

LOC4324891
Genomic Map