Assay Details
Gene Symbol: LOC4326950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780721.2 | XP_015636207.1 | 1 - 2 | 257 | 55 | |
GenBank mRNA | AK121373.1 | - | 1 - 2 | 188 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
