Assay Details
Gene Symbol: LOC4326950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780721.2 | XP_015636207.1 | 2 - 3 | 514 | 73 | |
GenBank mRNA | AK121373.1 | - | 2 - 3 | 445 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
