Assay Details
Gene Symbol: LOC4326957 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795467.2 | XP_015650953.1 | 10 - 11 | 1237 | 63 | |
GenBank mRNA | AK072309.1 | - | 10 - 11 | 1218 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
