Assay Details
Gene Symbol: LOC4326957 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795467.2 | XP_015650953.1 | 1 - 2 | 275 | 126 | |
GenBank mRNA | AK072309.1 | - | 1 - 2 | 256 | 126 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015795467.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
