Assay Details
Gene Symbol: LOC4325239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015793630.2 | XP_015649116.1 | 1 - 2 | 349 | 66 | |
GenBank mRNA | AK108183.1 | - | 1 - 2 | 323 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
