Assay Details
Gene Symbol: LOC4328203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771359.2 | XP_015626845.1 | 1 - 2 | 269 | 105 | |
GenBank mRNA | AK070078.1 | - | 1 - 2 | 270 | 105 | |
AK103414.1 | - | 1 - 2 | 215 | 105 | ||
AK106973.1 | - | 1 - 2 | 233 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771359.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
