Assay Details
Gene Symbol: LOC4328203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771354.2 | XP_015626840.1 | 3 - 4 | 594 | 68 | |
XM_015771355.2 | XP_015626841.1 | 3 - 4 | 594 | 68 | ||
XM_015771356.2 | XP_015626842.1 | 3 - 4 | 594 | 68 | ||
XM_015771359.2 | XP_015626845.1 | 3 - 4 | 686 | 68 | ||
GenBank mRNA | AK070078.1 | - | 3 - 4 | 687 | 68 | |
AK103414.1 | - | 3 - 4 | 632 | 68 | ||
AK106973.1 | - | 3 - 4 | 650 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
