Assay Details
Gene Symbol: LOC4328203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771354.2 | XP_015626840.1 | 5 - 6 | 845 | 144 | |
XM_015771355.2 | XP_015626841.1 | 5 - 6 | 845 | 144 | ||
XM_015771359.2 | XP_015626845.1 | 5 - 6 | 937 | 144 | ||
GenBank mRNA | AK070078.1 | - | 5 - 6 | 938 | 144 | |
AK103414.1 | - | 5 - 6 | 883 | 144 | ||
AK106973.1 | - | 5 - 6 | 901 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
