Assay Details
Gene Symbol: LOC4328203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771354.2 | XP_015626840.1 | 7 - 8 | 1077 | 129 | |
XM_015771356.2 | XP_015626842.1 | 6 - 7 | 939 | 129 | ||
XM_015771359.2 | XP_015626845.1 | 7 - 8 | 1169 | 129 | ||
GenBank mRNA | AK103414.1 | - | 7 - 8 | 1115 | 129 | |
AK106973.1 | - | 7 - 8 | 1133 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
