Assay Details
| Gene Symbol: LOC4328787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771884.2 | XP_015627370.1 | 11 - 12 | 1670 | 101 | |
| GenBank mRNA | AK288680.1 | - | 11 - 12 | 1632 | 101 | |
More Information
Set Membership: |
|

LOC4328787
Genomic Map