Assay Details
| Gene Symbol: LOC4346120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793203.2 | XP_015648689.1 | 2 - 3 | 470 | 132 | |
| GenBank mRNA | AK107138.1 | - | 2 - 3 | 456 | 132 | |
More Information
Set Membership: |
|

LOC4346120
Genomic Map