Assay Details
Gene Symbol: LOC4346967 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795666.2 | XP_015651152.1 | 6 - 7 | 1141 | 113 | |
GenBank mRNA | AK120537.1 | - | 6 - 7 | 1085 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
