Assay Details
Gene Symbol: LOC4346967 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795666.2 | XP_015651152.1 | 7 - 8 | 1323 | 145 | |
GenBank mRNA | AK120537.1 | - | 7 - 8 | 1267 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() |
