Assay Details
| Gene Symbol: LOC4348305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759391.2 | XP_015614877.1 | 5 - 6 | 657 | 120 | |
| GenBank mRNA | AK072694.1 | - | 5 - 6 | 605 | 120 | |
| KC140130.1 | - | 5 - 6 | 627 | 120 | ||
More Information
Set Membership: |
|

LOC4348305
Genomic Map