Assay Details
| Gene Symbol: LOC4349333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757426.2 | XP_015612912.1 | 10 - 11 | 1456 | 113 | |
| XM_015757428.2 | XP_015612914.1 | 10 - 11 | 1471 | 113 | ||
| XM_015757429.2 | XP_015612915.1 | 9 - 10 | 1306 | 113 | ||
| GenBank mRNA | AB277360.1 | - | 10 - 11 | 1415 | 113 | |
| AK069361.1 | - | 10 - 11 | 1420 | 113 | ||
More Information
Set Membership: |
|

LOC4349333
Genomic Map