Assay Details
| Gene Symbol: LOC4349410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757820.2 | XP_015613306.1 | 4 - 5 | 1256 | 70 | |
| GenBank mRNA | AK109667.1 | - | 4 - 5 | 1262 | 70 | |
More Information
Set Membership: |
|

LOC4349410
Genomic Map