Assay Details
Gene Symbol: LOC4351542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764175.2 | XP_015619661.1 | 8 - 9 | 4049 | 91 | |
XM_015764177.2 | XP_015619663.1 | 8 - 9 | 4040 | 91 | ||
GenBank mRNA | AK060480.1 | - | 5 - 6 | 1143 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
