Assay Details
| Gene Symbol: LOC4351964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763711.2 | XP_015619197.1 | 1 - 2 | 392 | 64 | |
| GenBank mRNA | AK063071.1 | - | 1 - 2 | 447 | 64 | |
More Information
Set Membership: |
|

LOC4351964
Genomic Map