Assay Details
Gene Symbol: LOC4351982 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015763228.2 | XP_015618714.1 | 1 - 2 | 374 | 64 | |
GenBank mRNA | AK070295.1 | - | 1 - 2 | 381 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
