Assay Details
Gene Symbol: LOC4351982 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015763228.2 | XP_015618714.1 | 8 - 9 | 1141 | 81 | |
GenBank mRNA | AK070295.1 | - | 8 - 9 | 1148 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
