Assay Details
| Gene Symbol: LOC9270871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763882.2 | XP_015619368.1 | 3 - 4 | 1672 | 114 | |
| GenBank mRNA | AK099690.1 | - | 3 - 4 | 1673 | 114 | |
More Information
Set Membership: |
|

LOC9270871
Genomic Map