Assay Details
| Gene Symbol: LOC9270871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763882.2 | XP_015619368.1 | 7 - 8 | 2482 | 126 | |
| GenBank mRNA | AK099690.1 | - | 7 - 8 | 2483 | 126 | |
| AK105377.1 | - | 1 - 2 | 122 | 126 | ||
More Information
Set Membership: |
|

LOC9270871
Genomic Map