Assay Details
| Gene Symbol: LOC9270871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763882.2 | XP_015619368.1 | 8 - 9 | 2613 | 87 | |
| GenBank mRNA | AK099690.1 | - | 8 - 9 | 2614 | 87 | |
| AK105377.1 | - | 2 - 3 | 253 | 87 | ||
More Information
Set Membership: |
|

LOC9270871
Genomic Map