Assay Details
| Gene Symbol: LOC4352411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765194.2 | XP_015620680.1 | 7 - 8 | 1281 | 66 | |
| XM_015765195.2 | XP_015620681.1 | 7 - 8 | 1272 | 66 | ||
| XM_015765197.2 | XP_015620683.1 | 7 - 8 | 1281 | 66 | ||
| XM_015765198.2 | XP_015620684.1 | 7 - 8 | 1272 | 66 | ||
| GenBank mRNA | AK058923.1 | - | 2 - 3 | 88 | 66 | |
More Information
Set Membership: |
|

LOC4352411
Genomic Map