Assay Details
Gene Symbol: LOC4352545 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015763552.2 | XP_015619038.1 | 1 - 2 | 176 | 77 | |
XM_015763553.2 | XP_015619039.1 | 1 - 2 | 176 | 77 | ||
GenBank mRNA | AK072828.1 | - | 1 - 2 | 177 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015763552.2, XM_015763553.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
