Assay Details
Gene Symbol: LOC4352701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765306.2 | XP_015620792.1 | 1 - 2 | 244 | 91 | |
GenBank mRNA | AK111887.1 | - | 1 - 2 | 201 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015765306.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
