Assay Details
Gene Symbol: LOC9269535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764715.2 | XP_015620201.1 | 5 - 6 | 852 | 101 | |
XM_015764716.2 | XP_015620202.1 | 5 - 6 | 898 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
