Assay Details
Gene Symbol: LOC9269535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764715.2 | XP_015620201.1 | 8 - 9 | 1513 | 125 | |
XM_015764716.2 | XP_015620202.1 | 8 - 9 | 1559 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
