Assay Details
| Gene Symbol: LOC4352843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763106.2 | XP_015618592.1 | 4 - 5 | 793 | 66 | |
| GenBank mRNA | AK059924.1 | - | 4 - 5 | 767 | 66 | |
More Information
Set Membership: |
|

LOC4352843
Genomic Map