Assay Details
Gene Symbol: LOC4350081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762399.1 | XP_015617885.1 | 10 - 11 | 1767 | 68 | |
XM_015762400.2 | XP_015617886.1 | 10 - 11 | 1757 | 68 | ||
GenBank mRNA | AK062044.1 | - | 5 - 6 | 975 | 68 | |
AK067129.1 | - | 10 - 11 | 1733 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
