Assay Details
Gene Symbol: LOC4350081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762399.1 | XP_015617885.1 | 14 - 15 | 2166 | 83 | |
XM_015762400.2 | XP_015617886.1 | 14 - 15 | 2156 | 83 | ||
GenBank mRNA | AK062044.1 | - | 9 - 10 | 1374 | 83 | |
AK067129.1 | - | 14 - 15 | 2132 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
