Assay Details
Gene Symbol: LOC4350081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762399.1 | XP_015617885.1 | 2 - 3 | 516 | 78 | |
XM_015762400.2 | XP_015617886.1 | 2 - 3 | 506 | 78 | ||
GenBank mRNA | AK067129.1 | - | 2 - 3 | 482 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
