Assay Details
Gene Symbol: LOC4350552 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761129.2 | 10 - 11 | 1296 | 104 | ||
GenBank mRNA | AK107085.1 | - | 4 - 5 | 348 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
