Assay Details
Gene Symbol: LOC4350552 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761129.2 | 11 - 12 | 1387 | 112 | ||
GenBank mRNA | AK107085.1 | - | 5 - 6 | 439 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
