Assay Details
| Gene Symbol: LOC4350674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760138.2 | XP_015615624.1 | 1 - 2 | 407 | 76 | |
| GenBank mRNA | AK072076.1 | - | 1 - 2 | 318 | 76 | |
More Information
Set Membership: |
|

LOC4350674
Genomic Map