Assay Details
Gene Symbol: LOC4350790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015760252.2 | XP_015615738.1 | 1 - 2 | 428 | 71 | |
GenBank mRNA | AK064672.1 | - | 1 - 2 | 428 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
