Assay Details
Gene Symbol: LOC4350790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015760252.2 | XP_015615738.1 | 2 - 3 | 466 | 86 | |
GenBank mRNA | AK064672.1 | - | 2 - 3 | 466 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
