Assay Details
| Gene Symbol: LOC4328609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771730.2 | XP_015627216.1 | 1 - 2 | 145 | 112 | |
| GenBank mRNA | AK067403.1 | - | 1 - 2 | 144 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771730.2 transcripts.
Set Membership: |
|

LOC4328609
Genomic Map