Assay Details
Gene Symbol: LOC4329595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769587.2 | XP_015625073.1 | 2 - 3 | 224 | 71 | |
GenBank mRNA | AK107058.1 | - | 2 - 3 | 230 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
